Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brilliant Crocein, Thermo Scientific Chemicals

Catalog Number p-7022049
Click to view available options
Quantity:
50 g
250 g
5413-75-2
C22H16N4Na2O7S2
558.491
MFCD00003907
BMBHMIGLJQAUTJ-HKWNQDJZSA-N
C.I. 27290; Crocein Scarlet MOO
88641610
(8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]
5413-75-2
C22H16N4Na2O7S2
558.491
MFCD00003907
BMBHMIGLJQAUTJ-HKWNQDJZSA-N
C.I. 27290; Crocein Scarlet MOO
88641610
(8Z)-7-oxo-8-[(4-phenyldiazenylphenyl)hydrazinylidene]naphthalene-1,3-disulfonic acid;sodium
C1=CC=C(C=C1)N=NC2=CC=C(C=C2)NN=C3C(=O)C=CC4=CC(=CC(=C43)S(=O)(=O)O)S(=O)(=O)O.[Na].[Na]

Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Brilliant Crocein is used mainly as a wool, silk, leather paper and leather dyeing. Can also be used for plastic, wood, spices, electrical-controlled aluminum, medicine color. Can make the ink, the heavy metal salt can be used as pigment.

Solubility
Soluble in water

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Solubility Information Soluble in water
Formula Weight 556.49
Quantity 50 g
Chemical Name or Material Brilliant Crocein
EINECSNumber 226-502-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit