Metrohm Electrolyte Solutions for Metrohm™ Karl Fischer Titrator Electrodes
Manufacturer: Metrohm SNGTI94443
 View more versions of this product
Catalog No. 20-094-443
  Check Availability
Add to Cart
    Specifications
    Applications Titration in non-aqueous solutions and reference electrolyte
    Volume (Metric) Sample 250mL
    Type Electrolyte Solution
    Filling Solution Type Lithium Chloride
    For Use With Metrohm™ Karl Fischer titrator electrode
  • Description & Specifications
Related Products
Product Content Feedback