KINEMATICA Polytron Dispersing Aggregates
For use with P2100 drive unit
Manufacturer: KINEMATICA 11030070
 View more versions of this product
Catalog No. 08-451-65
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Capacity (Metric) 2 to 250mL
    Type Generator
    For Use With (Application) Ideal for production of suspensions and emulsions
    For Use With (Equipment) Polytron™ PT 2100 homogenizer
    Material Stainless Steel
    Model PT DA 2120/2EC
  • Description & Specifications
Related Products
Product Content Feedback