KINEMATICA Polytron Dispersing Aggregates

For use with P2100 drive unit

Manufacturer: KINEMATICA 11030071

 View more versions of this product

Catalog No. 08-451-181

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Capacity (Metric) 100 to 2000mL
    Contains Rotor with knives
    Type Generator
    For Use With (Application) Ideal for processing of larger cell tissue samples
    For Use With (Equipment) Polytron™ PT 2100 homogenizer
    Material Stainless Steel
    Model PT DA 20/2MEC

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit