Metrohm Model 727 Stirring Stand
Manufacturer: Metrohm 020757078
 View more versions of this product
Catalog No. 20-757-078
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Titration stand and stirring accessory is available witn magnetic stirrer and electrode rinse or with propeller stirrer. Models for 9VDC connect directly to Metrohm Titrators.
  • Description & Specifications
Related Products
Product Content Feedback