Learn More
LIDOCAINE 50 MG
Na+ channel blocker
$64.00
Chemical Identifiers
CAS | 137-58-6 |
---|---|
Molecular Formula | C14H22N2O |
Molecular Weight (g/mol) | 234.343 |
InChI Key | NNJVILVZKWQKPM-UHFFFAOYSA-N |
Synonym | 2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, cappicaine, duncaine, esracaine, gravocain, lidocaine, lidoderm, lignocaine, xylocaine |
PubChem CID | 3676 |
ChEBI | CHEBI:6456 |
IUPAC Name | 2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide |
SMILES | CCN(CC)CC(=O)NC1=C(C=CC=C1C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
30-575-0
![]() |
Tocris Bioscience
3057/50 |
50mg |
Each for $64.00
|
|
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Chemical Identifiers
137-58-6 | |
234.343 | |
2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, cappicaine, duncaine, esracaine, gravocain, lidocaine, lidoderm, lignocaine, xylocaine | |
CHEBI:6456 | |
CCN(CC)CC(=O)NC1=C(C=CC=C1C)C |
C14H22N2O | |
NNJVILVZKWQKPM-UHFFFAOYSA-N | |
3676 | |
2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide |
Specifications
137-58-6 | |
2-diethylamino-n-2,6-dimethylphenyl acetamide, anestacon, cappicaine, duncaine, esracaine, gravocain, lidocaine, lidoderm, lignocaine, xylocaine | |
CCN(CC)CC(=O)NC1=C(C=CC=C1C)C | |
234.343 | |
CHEBI:6456 | |
>99% |
C14H22N2O | |
NNJVILVZKWQKPM-UHFFFAOYSA-N | |
2-(diethylamino)-N-(2,6-dimethylphenyl)acetamide | |
3676 | |
234.34 | |
Lidocaine |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.