Learn More
PicoStudio BristleBots
SCI_ED
This BristleBots pack includes everything needed to build BristleBots in your home or classroom to teach basic engineering, motors, circuits, and physics concepts (i.e. balance principles).
Manufacturer:  PicoStudio PS30091
 View more versions of this product
Catalog No. S4131922
Add to cart
Alert:
Small Parts - Not recommended for children under 3 years old.
Description
Description
  • A Bristlebot is a small robot made by combining a toothbrush head and a vibrating pager motor.
  • Vibrations from the motor travel down the bristles and cause the brush to move and spin on flat surfaces.
  • Adjust different parts by bending, twisting, and angling the chenille stems into different positions and see how it affects the movement and direction of the Bristlebot.
  • Build time: 3 to 10 minutes.
  • Suitable for ages 5+.
  • Ideal for camps, scout troops, homeschool, libraries, after-school, STEM/STEAM groups, birthday party, and classroom activities.
  • Race or battle them against each other.
Specifications
Specifications
20 Bristle Heads, 20 Vibration Motors (1.5 V), 20 Button Batteries (1.5 V), 40 Pipe Cleaners, 40 Google Eyes, 20 Double Sided Tape
STEM Kit
20 Pk.