Learn More
PicoStudio BristleBots Products for Science Education
This BristleBots pack includes everything needed to build BristleBots in your home or classroom to teach basic engineering, motors, circuits, and physics concepts (i.e. balance principles).
$19.99 - $89.99
Specifications
Type STEM Kit
View More Specs
Alert:
Small Parts - Not recommended for children under 3 years old.
Products 2
Catalog Number Mfr. No. Kit Size Includes Price Quantity & Availability  
Catalog Number Mfr. No. Kit Size Includes Price Quantity & Availability  
S4131923 Products for Science Education
View Documents
PicoStudio
PS30092
4 Pk. 4 Bristle Heads, 4 Vibration Motors (1.5 V), 4 Button Batteries (1.5 V), 8 Pipe Cleaners, 8 Google Eyes, 4 Double Sided Tape
Each for $19.99
Add to cart
S4131922 Products for Science Education
View Documents
PicoStudio
PS30091
20 Pk. 20 Bristle Heads, 20 Vibration Motors (1.5 V), 20 Button Batteries (1.5 V), 40 Pipe Cleaners, 40 Google Eyes, 20 Double Sided Tape
Each for $89.99
Add to cart
Description
Description
  • A Bristlebot is a small robot made by combining a toothbrush head and a vibrating pager motor.
  • Vibrations from the motor travel down the bristles and cause the brush to move and spin on flat surfaces.
  • Adjust different parts by bending, twisting, and angling the chenille stems into different positions and see how it affects the movement and direction of the Bristlebot.
  • Build time: 3 to 10 minutes.
  • Suitable for ages 5+.
  • Ideal for camps, scout troops, homeschool, libraries, after-school, STEM/STEAM groups, birthday party, and classroom activities.
  • Race or battle them against each other.
Specifications
Specifications
STEM Kit