Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRL 15572 hydrochloride, Tocris

Selective h5-HT1D antagonist

Manufacturer:  R&D Systems 1207/50

Quantity 50mg

 View more versions of this product

Catalog No. 12-075-0


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

>98%
443.42
BRL 15572 hydrochloride
C25H27ClN2O·HCl
3-[4-(4-Chlorophenyl)piperazin-1-yl]-1, 1-diphenyl-2-propanol hydrochloride

Specifications

50mg
>98%
443.42
BRL 15572 hydrochloride
1173022-77-9
C25H27ClN2O·HCl
3-[4-(4-Chlorophenyl)piperazin-1-yl]-1, 1-diphenyl-2-propanol hydrochloride

Safety and Handling

Recommended Storage : Desiccate at +4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit