Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BRL 37344, sodium salt, Tocris Bioscience™

Catalog Number 094810
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
127299-93-8
C19H21ClNNaO4
385.82
SNJIJYKMYQRHRC-QRGZVCNKSA-M
4-s-2-s-2-3-chlorophenyl-2-hydroxyethyl amino propyl phenoxy acetic acid sodium salt, sodium 2-4-2s-2-2s-2-3-chlorophenyl-2-hydroxyethyl amino propyl phenoxy acetate
91667707
sodium;2-[4-[(2R)-2-[[(2S)-2-(3-chlorophenyl)-2-hydroxyethyl]amino]propyl]phenoxy]acetate
CC(CC1=CC=C(C=C1)OCC(=O)[O-])NCC(C2=CC(=CC=C2)Cl)O.[Na+]
This item is not returnable. View return policy
127299-93-8
C19H21ClNNaO4
385.82
SNJIJYKMYQRHRC-QRGZVCNKSA-M
4-s-2-s-2-3-chlorophenyl-2-hydroxyethyl amino propyl phenoxy acetic acid sodium salt, sodium 2-4-2s-2-2s-2-3-chlorophenyl-2-hydroxyethyl amino propyl phenoxy acetate
91667707
sodium;2-[4-[(2R)-2-[[(2S)-2-(3-chlorophenyl)-2-hydroxyethyl]amino]propyl]phenoxy]acetate
CC(CC1=CC=C(C=C1)OCC(=O)[O-])NCC(C2=CC(=CC=C2)Cl)O.[Na+]

β3 agonist

Potent and selective β3 adrenoceptor agonist (Ki values are 287, 1750 and 1120 nM for β3, β1 and β2 receptors respectively).
Quantity 10 mg
Formula Weight 385.82
Percent Purity >98%
Chemical Name or Material BRL 37344, sodium salt
Recommended Storage Store at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit