Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BRL 44408 maleate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
BRL 44408 maleate is a selective α2A-adrenoceptor antagonist (Ki = 1.7 nM and 144.5 nM at α2A and α2B-adrenergic receptors respectively). Increases hippocampal noradrenalin release following systemic administration.
Specifications
Quantity | 10 mg |
Formula Weight | 331.37 |
Percent Purity | >98% |
Chemical Name or Material | BRL 44408 maleate |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction