Learn More
BRL 52537 hydrochloride, Tocris
Potent and selective κ agonist
Manufacturer:  R&D Systems 0699/10
Quantity 10mg
 View more versions of this product
Catalog No. 06-991-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
391.77
BRL 52537 hydrochloride
C18H24Cl2N2O·HCl
(±)-1-(3, 4-Dichlorophenyl)acetyl-2-(1-pyrrolidinyl)methylpiperidine hydrochloride
Specifications
10mg
>98%
391.77
BRL 52537 hydrochloride
112282-24-3
C18H24Cl2N2O·HCl
(±)-1-(3, 4-Dichlorophenyl)acetyl-2-(1-pyrrolidinyl)methylpiperidine hydrochloride
Safety and Handling
Recommended Storage : Store at Room Temperature