Learn More
Brn3a Rabbit anti-Human,Mouse,Rat, KHL, Clone: 4L5, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma ZRB109825UL
 View more versions of this product
Catalog No. ZRB109825MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Brn3a Monoclonal antibody specifically detects Brn3a in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications
Specifications
Brn3a
4L5
KHL
Purified
POU4F1, BRN3A, RDC1
KLH-conjugted linear peptide corresponding to 19 amino acids from the internal region of human homeobox/POU domain protein 3A (Brn-3a).
∼48 kDa observed; 42.70 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
25 μL
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Monoclonal
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
30 μg/mL after reconstitution at 25 μL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
NP_006228.3
POU domain;class 4;transcription factor 1;Brain-specific homeobox/POU domain protein 3A;Brain-3A;Homeobox/POU domain protein RDC-1;Oct-T1
Rabbit
IgG
Protein A Purified
Epigenetics and Nuclear Function
Primary
Human, Mouse, Rat