Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brn3a Rabbit anti-Human,Mouse,Rat, KHL, Clone: 4L5, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Brn3a
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB109825MI
View Documents
MilliporeSigma
ZRB109825UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB10984XMI
View Documents
MilliporeSigma
ZRB10984X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Brn3a Monoclonal antibody specifically detects Brn3a in Human,Mouse,Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications

Specifications

Brn3a
KHL
Rabbit
POU domain;class 4;transcription factor 1;Brain-specific homeobox/POU domain protein 3A;Brain-3A;Homeobox/POU domain protein RDC-1;Oct-T1
KLH-conjugted linear peptide corresponding to 19 amino acids from the internal region of human homeobox/POU domain protein 3A (Brn-3a).
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Purified
Human, Mouse, Rat
POU4F1, BRN3A, RDC1
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit