Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromhexine Hydrochloride, BP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
500 g
1 kg
Packaging:
Amber Glass Bottle
611-75-6
C14H21Br2ClN2
412.59
SXIVGYTWSVJOLM-UHFFFAOYNA-N
hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
[H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N
611-75-6
C14H21Br2ClN2
412.59
SXIVGYTWSVJOLM-UHFFFAOYNA-N
hydrogen 2-(1-amino-1-cyclohexylethyl)-4,6-dibromoaniline chloride
[H+].[Cl-].CC(N)(C1CCCCC1)C1=CC(Br)=CC(Br)=C1N

B1552, 611-75-6, C14H20Br2N2•HCl

Spectrum™ Bromhexine Hydrochloride, BP is a mucolytic (expectorant) agent used in the treatment of respiratory disorders. Spectrum Chemical BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities..
Loss on Drying 1%
Sulfated Ash 0.1%
Quantity 1 kg
Packaging Amber Glass Bottle
CAS Max % 100%
Percent Purity 98.5-101.5%