Learn More
Brominated Vegetable Oil, FCC, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Drum
Steel Can
Quantity:
500 mL
V1031, 8016-94-2
Spectrum™ Brominated Vegetable Oil, FCC , also known as BVO, is a mix of plant-derived triglycerides and is used to help emulsify citrus flavored drinks. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Steel Can