Learn More
Brominated Wang polymer resin, 1% crossl. with DVB, 0.5-1.0mmol Br/g, 200-400 mesh, Acros Organics
Manufacturer:  Acros Organics 349230010
 View more versions of this product
Catalog No. AC349230010
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
for solid support synthesis
Specifications
Specifications
crosslinking: 1% DVB
0.5 to 1mmol/g
1g
MFCD00163168
Brominated Wang polymer resin
200 to 400 mesh
Brown or White to Yellow
Beads