Learn More
Brominated Wang polymer resin, 1% crossl. with DVB, 0.8-1.2mmol Br/g, 100-200 mesh, Acros Organics
Manufacturer:  Acros Organics 349220010
 View more versions of this product
Catalog No. AC349220010
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
for solid support synthesis
Specifications
Specifications
crosslinking: 1% DVB
0.8 to 1.2mmol/g
1g
MFCD00163168
Brominated Wang polymer resin
100 to 200 mesh
Brown or White to Yellow
Beads