Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brominated Wang polymer resin, 1% crossl. with DVB, 0.8-1.2mmol Br/g, 100-200 mesh, ACROS Organics™

Specifications

Chemical Name or Material Brominated Wang polymer resin
MDL Number MFCD00163168
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC349220010
View Documents
Acros Organics
349220010
1g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
AC349220050
View Documents
Acros Organics
349220050
5g This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

for solid support synthesis
Specifications

Specifications

Brominated Wang polymer resin
MFCD00163168
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit