Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromine-1,4-dioxane complex, 0.25M solution in 1,4-dioxane, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass bottle
15481-39-7,123-91-1
C4H8BrO2
168.01
MFCD00034818
GSQACUNMFGRAKY-UHFFFAOYSA-N
p-dioxane, compd. with bromine 1:1
57348859
bromine;1,4-dioxane
C1COCCO1.[Br]
15481-39-7,123-91-1
C4H8BrO2
168.01
MFCD00034818
GSQACUNMFGRAKY-UHFFFAOYSA-N
p-dioxane, compd. with bromine 1:1
57348859
bromine;1,4-dioxane
C1COCCO1.[Br]
Concentration 0.2 to 0.3M
Formula Weight 247.91
Physical Form Liquid
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: soluble.
Color Red
Quantity 100mL
Flash Point 17°C
Chemical Name or Material Bromine-1, 4-dioxane complex, 0.25M solution in 1, 4-dioxane

GHS H Statement
Suspected of causing cancer. Fatal if inhaled. May cause respiratory irritation. Toxic to aquatic life with long lasting effects. Causes serious eye irritation. Causes skin irritation. Highly flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Use personal protective equipment as required. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Avoid breathing dust/fume/gas/mist/vapors/spray. Avoid release to the environment. Keep container tightly closed. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Ground/Bond container and receiving equipment.

Danger