Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromine, 1M solution in acetic acid, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
7726-95-6,64-19-7
Br2
159.808
MFCD00010896
GDTBXPJZTBHREO-UHFFFAOYSA-N
bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish
24408
CHEBI:29224
molecular bromine
BrBr
7726-95-6,64-19-7
Br2
159.808
MFCD00010896
GDTBXPJZTBHREO-UHFFFAOYSA-N
bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish
24408
CHEBI:29224
molecular bromine
BrBr
C-X bond formation reagent
Linear Formula Br2
Concentration 0.90 to 1.10M
Density 3.111
Merck Index 15, 1401
Formula Weight 159.82
Physical Form Liquid
Packaging AcroSeal™ Glass Bottle
Solubility Solubility in water: soluble.
Color Red
Quantity 800mL
Flash Point 40°C
Chemical Name or Material Bromine, 1M solution in acetic acid
Show More Show Less

GHS P Statement Fatal if inhaled. Causes severe skin burns and eye damage. Very toxic to aquatic life. Flammable liquid and vapor.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Avoid release to the environment.

GHS Signal Word: Danger