Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromine, 99.6%, for analysis, ACROS Organics™

Click to view available options
Quantity:
25mL
250mL
1L
7726-95-6
Br2
159.82
MFCD00010896
GDTBXPJZTBHREO-UHFFFAOYSA-N
bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish
24408
CHEBI:29224
molecular bromine
BrBr
7726-95-6
Br2
159.82
MFCD00010896
GDTBXPJZTBHREO-UHFFFAOYSA-N
bromine,dibromine,brom,bromine solution,brome,bromo,broom,bromine water,bromo italian,bromo spanish
24408
CHEBI:29224
molecular bromine
BrBr
C-X bond formation reagent for analysis
Assay Percent Range 99.5% min. (Iodometry)
Linear Formula Br2
Fieser 03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47
Merck Index 15, 1401
Solubility Information Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2
Formula Weight 159.82
Percent Purity 99.6%
Grade Analysis
Density 3.1110g/mL
Packaging Glass bottle
Solubility Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2
Specific Gravity 3.111
Viscosity 0.314 mm2/s (25°C)
Boiling Point 58.7°C
Melting Point -7.2°C
Quantity 1L
Chemical Name or Material Bromine, p.a.
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 231-778-1

RTECSNumber : EF9100000

TSCA : TSCA