Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromine, 99+%, extra pure, Thermo Scientific Chemicals

Catalog Number AC402845000
Click to view available options
Quantity:
500 mL
Packaging:
Plastic Coated Glass Bottle
7726-95-6
Br
79.91
MFCD00010896
CPELXLSAUQHCOX-UHFFFAOYSA-M
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
24408
CHEBI:29224
[Br-]
7726-95-6
Br
79.91
MFCD00010896
CPELXLSAUQHCOX-UHFFFAOYSA-M
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
24408
CHEBI:29224
[Br-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 58.7°C
Melting Point -7.2°C
Color Red to Brown
Physical Form Fuming Liquid
Quantity 500 mL
Assay Percent Range 99% min. (Iodometry)
Linear Formula Br2
Fieser 03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47
Merck Index 15, 1401
Solubility Information Solubility in water: 35g/L (20°C). Other solubilities: soluble in concd hcl, aq solns of bromides, freely soluble in alcohol, ether, chcl3, ccl4, cs2
IUPAC Name bromide
Formula Weight 159.82
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic Coated Glass Bottle
Specific Gravity 3.111
Density 3.1110g/mL
Chemical Name or Material Bromine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Fatal if inhaled.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 231-778-1

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit