Learn More
Bromine, 1M solution in trimethyl phosphate, AcroSeal™, Thermo Scientific Chemicals
$8.99
Specifications
Chemical Name or Material | Bromine |
---|---|
Name Note | 1M Solution in trimethyl Phosphate |
CAS | 7726-95-6, 512-56-1 |
Molecular Formula | Br |
Linear Formula | Br2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC428721000
|
Thermo Scientific Chemicals
428721000 |
100 mL | AcroSeal™ Glass Bottle |
Each
for $8.99
Save 89% Reg :
Order in Bulk
Request bulk or custom formats |
|
||||
AC428728000
|
Thermo Scientific Chemicals
428728000 |
800 mL | AcroSeal™ Glass bottle | Item Discontinued Due to product restrictions, please Sign In to purchase or view availability for this product.This item has been discontinued by the supplier. | N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Bromine | |
7726-95-6, 512-56-1 | |
Br2 | |
15,1401 | |
CPELXLSAUQHCOX-UHFFFAOYSA-M | |
79.91 | |
CHEBI:29224 | |
ACS Reagent | |
1.3070g/mL |
1M Solution in trimethyl Phosphate | |
Br | |
113°C | |
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish | |
[Br-] | |
24408 | |
159.8 | |
1.307 |
Safety and Handling
GHS H Statement
May cause genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
GHS Signal Word: Danger
EINECSNumber : 231-778-1
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.