Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Bromo-2'-acetonaphthone 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
613-54-7
C12H9BrO
249.107
MFCD00004109
YHXHHGDUANVQHE-UHFFFAOYSA-N
2-bromo-2'-acetonaphthone,bromomethyl 2-naphthyl ketone,2-bromoacetyl naphthalene,2-bromo-1-naphthalen-2-yl ethanone,2-bromo-1-2-naphthyl-1-ethanone,ethanone, 2-bromo-1-2-naphthalenyl,2-bromo-1-2-naphthyl ethanone,2-bromo-1-naphthalen-2-yl ethan-1-one,alpha-bromo-2'-acetonaphthone,2-bromo-1-naphthalen-2-yl-ethanone
69179
2-bromo-1-naphthalen-2-ylethanone
C1=CC=C2C=C(C=CC2=C1)C(=O)CBr
613-54-7
C12H9BrO
249.107
MFCD00004109
YHXHHGDUANVQHE-UHFFFAOYSA-N
2-bromo-2'-acetonaphthone,bromomethyl 2-naphthyl ketone,2-bromoacetyl naphthalene,2-bromo-1-naphthalen-2-yl ethanone,2-bromo-1-2-naphthyl-1-ethanone,ethanone, 2-bromo-1-2-naphthalenyl,2-bromo-1-2-naphthyl ethanone,2-bromo-1-naphthalen-2-yl ethan-1-one,alpha-bromo-2'-acetonaphthone,2-bromo-1-naphthalen-2-yl-ethanone
69179
2-bromo-1-naphthalen-2-ylethanone
C1=CC=C2C=C(C=CC2=C1)C(=O)CBr
Color Brown-Purple
Infrared Spectrum Authentic
Quantity 50g
Packaging Glass bottle
Melting Point 82°C to 85°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C10H7COCH2Br
Formula Weight 249.11
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material α-Bromo-2'-acetonaphthone, 98%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 210-348-7

TSCA : TSCA