Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoacetaldehyde diethyl acetal, 97%, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
2032-35-1
C6H13BrO2
197.07
MFCD00000214
LILXDMFJXYAKMK-UHFFFAOYSA-N
bromoacetaldehyde diethyl acetal,bromoacetal,2,2-diethoxyethyl bromide,ethane, 2-bromo-1,1-diethoxy,bromacetal,1-bromo-2,2-diethoxyethane,diethyl bromoacetal,2-bromoacetaldehyde diethyl acetal,1,1-diethoxy-2-bromoethane,diethyl bromoacetaldehyde acetal
74852
2-bromo-1,1-diethoxyethane
CCOC(CBr)OCC
2032-35-1
C6H13BrO2
197.07
MFCD00000214
LILXDMFJXYAKMK-UHFFFAOYSA-N
bromoacetaldehyde diethyl acetal,bromoacetal,2,2-diethoxyethyl bromide,ethane, 2-bromo-1,1-diethoxy,bromacetal,1-bromo-2,2-diethoxyethane,diethyl bromoacetal,2-bromoacetaldehyde diethyl acetal,1,1-diethoxy-2-bromoethane,diethyl bromoacetaldehyde acetal
74852
2-bromo-1,1-diethoxyethane
CCOC(CBr)OCC
Packaging Glass bottle
Specific Gravity 1.28
Boiling Point 66.0°C to 67.0°C (18.0mmHg)
Flash Point 61°C
Quantity 100mL
Refractive Index 1.4380 to 1.4400
Density 1.2800g/mL
Assay Percent Range 97%
Linear Formula BrCH2CH(OC2H5)2
Beilstein 01,625
Solubility Information Solubility in water: immiscible
Formula Weight 197.07
Percent Purity 97%
Chemical Name or Material Bromoacetaldehyde diethyl acetal, 97%
Show More Show Less

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Fatal if inhaled.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.

GHS Signal Word: Danger

EINECSNumber : 217-989-1

TSCA : TSCA