Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoacetyl chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
22118-09-8
C2H2BrClO
157.39
MFCD00000724
SYZRZLUNWVNNNV-UHFFFAOYSA-N
bromoacetyl chloride, acetyl chloride, bromo, bromo-acetyl chloride, monobromoacetic acid chloride, bromoacetylchloride, bromacetylchioride, bromacetylchloride, bromoacetylchioride, bromacetyl chloride, bromo-acetylchloride
89602
2-bromoacetyl chloride
C(C(=O)Cl)Br
22118-09-8
C2H2BrClO
157.39
MFCD00000724
SYZRZLUNWVNNNV-UHFFFAOYSA-N
bromoacetyl chloride, acetyl chloride, bromo, bromo-acetyl chloride, monobromoacetic acid chloride, bromoacetylchloride, bromacetylchioride, bromacetylchloride, bromoacetylchioride, bromacetyl chloride, bromo-acetylchloride
89602
2-bromoacetyl chloride
C(C(=O)Cl)Br
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.9000g/mL
Color Yellow to Brown
Specific Gravity 1.9
Refractive Index 1.496
Boiling Point 127.0°C to 128.0°C
Quantity 25g
Assay Percent Range 94% min. (GC)
Linear Formula BrCH2COCl
Beilstein 02,215
Solubility Information Solubility in water: reacts
Formula Weight 157.39
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Bromoacetyl chloride
Show More Show Less

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 244-790-7

Recommended Storage : 4°C