Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromoacetyl chloride, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
22118-09-8
C2H2BrClO
157.39
MFCD00000724
SYZRZLUNWVNNNV-UHFFFAOYSA-N
bromoacetyl chloride,acetyl chloride, bromo,bromo-acetyl chloride,monobromoacetic acid chloride,bromoacetylchloride,bromacetylchioride,bromacetylchloride,bromoacetylchioride,bromacetyl chloride,bromo-acetylchloride
89602
2-bromoacetyl chloride
C(C(=O)Cl)Br
22118-09-8
C2H2BrClO
157.39
MFCD00000724
SYZRZLUNWVNNNV-UHFFFAOYSA-N
bromoacetyl chloride,acetyl chloride, bromo,bromo-acetyl chloride,monobromoacetic acid chloride,bromoacetylchloride,bromacetylchioride,bromacetylchloride,bromoacetylchioride,bromacetyl chloride,bromo-acetylchloride
89602
2-bromoacetyl chloride
C(C(=O)Cl)Br
Density 1.9000g/mL
Infrared Spectrum Authentic
Quantity 500g
Refractive Index 1.496
Color Yellow to Brown
Packaging Glass bottle
Boiling Point 127.0°C to 128.0°C
Specific Gravity 1.9
Assay Percent Range 94% min. (GC)
Linear Formula BrCH2COCl
Beilstein 02,215
Solubility Information Solubility in water: reacts
Formula Weight 157.39
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Bromoacetyl chloride
Show More Show Less

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 244-790-7

Recommended Storage : 4°C