Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromobenzene-d5, for NMR, 99+% atom D, ACROS Organics™

Click to view available options
Quantity:
5mL
Packaging:
Glass bottle
4165-57-5
C6H5Br
162.041
QARVLSVVCXYDNA-RALIUCGRSA-N
bromobenzene-d5,bromo 2h5 benzene,1-bromo 2 h? benzene,bromobenzene d5,benzene-d5, bromo,1-bromo 2h5 benzene,c6d5br,bromobenzene-d5, 99.5 atom % d,bromobenzene-d5, reagent grade, >=99 atom % d cp
2723968
1-bromo-2,3,4,5,6-pentadeuteriobenzene
C1=CC=C(C=C1)Br
4165-57-5
C6H5Br
162.041
QARVLSVVCXYDNA-RALIUCGRSA-N
bromobenzene-d5,bromo 2h5 benzene,1-bromo 2 h? benzene,bromobenzene d5,benzene-d5, bromo,1-bromo 2h5 benzene,c6d5br,bromobenzene-d5, 99.5 atom % d,bromobenzene-d5, reagent grade, >=99 atom % d cp
2723968
1-bromo-2,3,4,5,6-pentadeuteriobenzene
C1=CC=C(C=C1)Br
Color Colorless
Deuterium (D) 99.0% min.
Packaging Glass bottle
Water 0.1% max. (Coulometric)
Quantity 5mL
Linear Formula C6D5Br
Beilstein 05, III, 561
Formula Weight 162.06
Physical Form Liquid
Grade NMR
Chemical Name or Material Bromobenzene-d5, 99+ atom % D
Show More Show Less

GHS H Statement:
Causes skin irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.

GHS P Statement:
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning