Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromochlorophenol Blue, sodium salt, pure, water soluble, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
102185-52-4
C19H9Br2Cl2O5S-
580.044
MFCD00001612
KGDDIJKLMLNSCH-LICLKQGHSA-M
131698806
2-[(Z)-(3-bromo-5-chloro-4-hydroxyphenyl)-(3-bromo-5-chloro-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Cl)C3=CC(=C(C(=C3)Br)O)Cl)S(=O)(=O)[O-]
102185-52-4
C19H9Br2Cl2O5S-
580.044
MFCD00001612
KGDDIJKLMLNSCH-LICLKQGHSA-M
131698806
2-[(Z)-(3-bromo-5-chloro-4-hydroxyphenyl)-(3-bromo-5-chloro-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]benzenesulfonate
C1=CC=C(C(=C1)C(=C2C=C(C(=O)C(=C2)Br)Cl)C3=CC(=C(C(=C3)Br)O)Cl)S(=O)(=O)[O-]
Formula Weight 603.03
Grade Pure
Packaging Glass bottle
Quantity 5g
Chemical Name or Material Bromochlorophenol Blue, sodium salt