Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
25g
5g
Packaging:
Wide Mouth Amber Glass Bottle

Chemical Identifiers

76-60-8
C21H14Br4O5S
698.014
FRPHFZCDPYBUAU-UHFFFAOYSA-N
6451

High quality chemicals for laboratory and specialized industrial use

CAS 76-60-8
Molecular Formula C21H14Br4O5S
InChI Key FRPHFZCDPYBUAU-UHFFFAOYSA-N
SMILES CC1=C(C(=C(C=C1C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4C)Br)O)Br)Br)O)Br
IUPAC Name 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-3-methylphenol
Molecular Weight (g/mol) 698.014
PubChem CID 6451
Formula Weight 698.02
Packaging Wide Mouth Amber Glass Bottle
Quantity 5g
Chemical Name or Material Bromocresol Green
Grade Baker Analyzed™ ACS Reagent
Show More Show Less