Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromocresol Green Indicator, 0.04% w/v, Reagents
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass Bottle
Poly Bottle
Specifications
Specifications
Chemical Name or Material | Bromocresol Green Indicator, 0.04% w/v |
CAS | 7732-18-5, 62625-32-5 |
Boiling Point | 100°C |
Synonym | Bromocresol blue, Bromcresol Green, Bromcresol Blue |
Percent Purity | 0.04% |
Grade | Reagent |
Packaging | Glass Bottle |
Specific Gravity | 1.001 |
Color | Dark Blue |
Quantity | 100 mL |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction