Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green Indicator Aqueous Solution, 1% (w/v), Ricca Chemical

$165.80 - $192.10

Specifications

Physical Form Solution
Assay Percent Range 1% (w/v)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
R1209000500
View Documents
Ricca Chemical Company
R1209000500A
500mL Poly natural
Each for $165.80
Add to cart
 
R12090001A
View Documents
Ricca Chemical Company
R12090001A
1L Poly natural
Each for $192.10
Add to cart
 
Description

Description

Specifications

Specifications

Solution
1% (w/v)
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit