Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromocresol Green, Methyl Red Indicator, APHA for Alkalinity, Alcoholic, Reagents
Click to view available options
Quantity:
125 mL
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | Bromocresol Green - Methyl Red Indicator APHA for Alkalinity, Alcoholic |
Boiling Point | 72°C |
CAS | 64-17-5, 67-63-0, 67-63-0 |
Synonym | Bromcresol Green-Methyl Red, MRBG |
Grade | Reagent |
Specific Gravity | 0.7907 |
Quantity | 1 L |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction