Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Green Sodium Salt, ACS, Affymetrix/USB™

Click to view available options
Quantity:
5g
25g
Packaging:
Plastic bottle
62625-32-5
C21H13Br4O5SNa
MFCD00148898
62625-32-5
C21H13Br4O5SNa
MFCD00148898

Used as tracking dye in alkaline agarose gel electrophoresis

Formula Weight 720.00
Physical Form Powder
Grade ACS
Packaging Plastic bottle
Melting Point ~230°C (decomposition)
Quantity 25g
Chemical Name or Material Bromocresol Green sodium salt