Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Purple, BAKER™, J.T.Baker™

Click to view available options
Quantity:
5g
10g
Packaging:
Wide Mouth Amber Glass Bottle
115-40-2
C21H16Br2O5S
540.222
ABIUHPWEYMSGSR-UHFFFAOYSA-N
8273
CHEBI:86154
2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol
CC1=C(C(=CC(=C1)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)C)Br)O
115-40-2
C21H16Br2O5S
540.222
ABIUHPWEYMSGSR-UHFFFAOYSA-N
8273
CHEBI:86154
2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol
CC1=C(C(=CC(=C1)C2(C3=CC=CC=C3S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)C)Br)O

A grade of chemicals for laboratory use

Formula Weight 540.22
Packaging Wide Mouth Amber Glass Bottle
Quantity 5g
Chemical Name or Material Bromocresol Purple