Alfa Aesar™ Bromocresol Purple sodium salt, 0.04% w/v aq. soln.

Manufacturer: Alfa Aesar™ 38700M0

Quantity 6 x 100mL

 View more versions of this product

Catalog No. AA38700M0

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62625-30-3
    Formula Weight 562.22
    MDL Number MFCD00148896
    Synonym Bromocresol Purple, water soluble
    Chemical Name or Material Bromocresol Purple sodium salt

    Specifications

    Formula Weight 562.22
    Quantity 6 x 100mL
    Physical Form Liquid
    MDL Number MFCD00148896
    CAS 62625-30-3
    Synonym Bromocresol Purple, water soluble
    TSCA TSCA
    Concentration 0.04% w/v aq. soln.
    Color Orange
    Chemical Name or Material Bromocresol Purple sodium salt

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit