Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Purple, Sultone, ACS, Spectrum™

Click to view available options
Quantity:
5 g
25 g
1 kg
Packaging:
Amber Glass Bottle
115-40-2
115-40-2
CAS Max % 1
Grade ACS
Packaging Amber Glass Bottle
Quantity 25 g

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.