Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocresol Purple TS, (U.S.P. Test Solution), Spectrum™ Chemical
SDP

Catalog Number 18601096
Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
62625-30-3, 7732-18-5
C21H15Br2O5S
539.21
HJZFRHKAXRLQSG-XMHGGMMESA-M
2-[(3-bromo-4-hydroxy-5-methylphenyl)[(1E)-3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
CC1=CC(=CC(Br)=C1O)C(=C1\C=C(C)C(=O)C(Br)=C1)\C1=CC=CC=C1S([O-])(=O)=O
62625-30-3, 7732-18-5
C21H15Br2O5S
539.21
HJZFRHKAXRLQSG-XMHGGMMESA-M
2-[(3-bromo-4-hydroxy-5-methylphenyl)[(1E)-3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
CC1=CC(=CC(Br)=C1O)C(=C1\C=C(C)C(=O)C(Br)=C1)\C1=CC=CC=C1S([O-])(=O)=O

B-327, 62625-30-3

Spectrum™ Chemical Bromocresol Purple TS, (U.S.P. Test Solution) is a solution, sometimes designated with "TS or Test Solution", that is prepared according to the specifications of the USP Pharmacopeial Convention, for the purpose of conducting USP and National Formulary tests and assays.
CAS Max % 0.1%, 49.9%
Grade Chemical Solution
Packaging Amber Glass Bottle
Quantity 1 L
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit