Alfa Aesar™ Bromocyclohexane, 99%

Manufacturer: Alfa Aesar™ 3103122

Quantity 100g

 View more versions of this product

Catalog No. AA3103122

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-85-0
    Molecular Formula C6H11Br
    Formula Weight 163.06
    MDL Number MFCD00003819
    Beilstein 1098373
    Merck Index 14,2730
    Synonym Cyclohexyl bromide
    Chemical Name or Material Bromocyclohexane

    Specifications

    Formula Weight 163.06
    Assay Percent Range 99%
    Molecular Formula C6H11Br
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Beilstein 1098373
    MDL Number MFCD00003819
    Synonym Cyclohexyl bromide
    Melting Point −57°C
    Boiling Point 166° to 167°C
    Flash Point 62° (143°F)
    Sensitivity Air & Light Sensitive
    Refractive Index 1.4950
    Physical Form Liquid
    Density 1.335g/mL
    CAS 108-85-0
    Merck Index 14,2730
    Health Hazard 1 Warning
    TSCA TSCA
    Quantity 100g
    Health Hazard 2 GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    RTECS Number GU7171700
    Chemical Name or Material Bromocyclohexane
    Assay 99%

    GHS H Statement
    H315-H319-H335-H227
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit