Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromocyclohexane, 99%

Manufacturer:  Alfa Aesar 3103136

Quantity 500g

 View more versions of this product

Catalog No. AA3103136

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

108-85-0
163.06
1098373
Cyclohexyl bromide
C6H11Br
MFCD00003819
14,2730
Bromocyclohexane

Specifications

108-85-0
99%
163.06
MFCD00003819
14,2730
500g
99%
Liquid
C6H11Br
1.335 g/mL
1098373
Cyclohexyl bromide
Bromocyclohexane

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335-H227

GHS P Statement
P210-P261-P280-P305+P351+P338-P304+P340-P362-P312-P405-P403+P233-P501a

Warning

RTECSNumber : GU7171700

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit