Alfa Aesar™ Bromocyclooctane, 97%

Manufacturer: Alfa Aesar™ B2483922

Quantity 100g

 View more versions of this product

Catalog No. AAB2483922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1556-09-8
    Molecular Formula C8H15Br
    Formula Weight 191.12
    MDL Number MFCD00039603
    Beilstein 1901417
    Synonym Cyclooctyl bromide
    Chemical Name or Material Bromocyclooctane

    Specifications

    Formula Weight 191.12
    Molecular Formula C8H15Br
    MDL Number MFCD00039603
    CAS 1556-09-8
    Beilstein 1901417
    Synonym Cyclooctyl bromide
    Boiling Point 102° to 103°C (18mmHg)
    Quantity 100g
    Chemical Name or Material Bromocyclooctane
    Assay 97%

    Description

    Applications
    Bromocyclooctane exhibits antibacterial and antineoplastic properties. It is used as pharmaceutical intermediates.

    Notes
    Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit