Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bromocyclooctane, 97%

Click to view available options
Quantity:
5g
1556-09-8
C8H15Br
191.112
MFCD00039603
KFKLBMQLKLKHLU-UHFFFAOYSA-N
cyclooctyl bromide,cyclooctane, bromo,1-bromocyclooctane,cyclooctane,bromo,acmc-1bxzd,kfklbmqlklkhlu-uhfffaoysa
73783
bromocyclooctane
C1CCCC(CCC1)Br
1556-09-8
C8H15Br
191.112
MFCD00039603
KFKLBMQLKLKHLU-UHFFFAOYSA-N
cyclooctyl bromide,cyclooctane, bromo,1-bromocyclooctane,cyclooctane,bromo,acmc-1bxzd,kfklbmqlklkhlu-uhfffaoysa
73783
bromocyclooctane
C1CCCC(CCC1)Br
Applications
Bromocyclooctane exhibits antibacterial and antineoplastic properties. It is used as pharmaceutical intermediates.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Boiling Point 102°C to 103°C (18mmHg)
Quantity 5g
Beilstein 1901417
Formula Weight 191.12
Percent Purity 97%
Chemical Name or Material Bromocyclooctane