Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromodeoxyuridine/BrdU Rabbit anti-All, Clone: BRD/1539R, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP30731720UG
Description
Bromodeoxyuridine/BrdU Monoclonal specifically detects Bromodeoxyuridine/BrdU in All Species samples. It is validated for ELISA.Specifications
Bromodeoxyuridine/BrdU | |
Monoclonal | |
0.2 mg/ml | |
ELISA | |
Bromodeoxyuridine, BUdr | |
Bromodeoxyuridine/BrdU conjugated to KLH | |
20 μg | |
Primary | |
Store at 4°C. | |
IgG |
ELISA | |
BRD/1539R | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
All | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title