Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodeoxyuridine/BrdU Rabbit anti-All, Clone: BRD/1539R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30731710UG

 View more versions of this product

Catalog No. NB122107


Add to cart

Description

Description

Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in All samples. It is validated for ELISA
Specifications

Specifications

Bromodeoxyuridine/BrdU
BRD/1539R
Unconjugated
10 mM PBS with 0.05% BSA
Bromodeoxyuridine, BUdr
Bromodeoxyuridine/BrdU conjugated to KLH
Protein A or G purified
RUO
Primary
All
ELISA
0.2 mg/ml
ELISA
Purified
Rabbit
IgG
100 μg
Store at 4°C.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit