Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromodeoxyuridine/BrdU Rabbit anti-All, Clone: BRD/1539R, Novus Biologicals™

Rabbit Monoclonal Antibody

$159.82 - $406.52

Specifications

Antigen Bromodeoxyuridine/BrdU
Clone BRD/1539R
Concentration 0.2 mg/ml
Dilution ELISA
Applications ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122108
View Documents
Novus Biologicals
NBP30731720UG
20 μg
Each for $159.82
Add to cart
 
NB122107
View Documents
Novus Biologicals
NBP30731710UG
100 μg
Each for $406.52
Add to cart
 
Description

Description

Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in All samples. It is validated for ELISA
Specifications

Specifications

Bromodeoxyuridine/BrdU
0.2 mg/ml
ELISA
Purified
RUO
10 mM PBS with 0.05% BSA
Bromodeoxyuridine/BrdU conjugated to KLH
Monoclonal
Protein A or G purified
BRD/1539R
ELISA
Unconjugated
Rabbit
All
Bromodeoxyuridine, BUdr
IgG
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit