Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bromodichloromethane, 98%, stab. with potassium carbonate

Manufacturer:  Alfa Aesar™ A1129218

Quantity 50g

 View more versions of this product

Catalog No. AAA1129218

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

75-27-4
163.83
1697005
FC-20B1
BrCHCl2
MFCD00000824
14,1417
Bromodichloromethane, Stabilized with potassium carbonate

Specifications

75-27-4
163.83
−57°C
14,1417
87° to 89°C
Chloroform-like
98%
BrCHCl2
MFCD00000824
1697005
FC-20B1
50g
Bromodichloromethane, Stabilized with potassium carbonate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H350-H302-H315-H319-H335
May cause cancer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : PA5310000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit