Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bromodiphenylmethane, 90%, Acros Organics
Click to view available options
Quantity:
100g
500g
Specifications
Melting Point | 33.0°C to 42.0°C |
Packaging | Glass bottle |
Boiling Point | 184.0°C (20.0 mmHg) |
Quantity | 100g |
Flash Point | >110°C |
Assay Percent Range | 90% |
Linear Formula | (C6H5)2CHBr |
Beilstein | 05,592 |
Solubility Information | Solubility in water: insoluble |
Formula Weight | 247.13 |
Show More |
Safety and Handling
EINECSNumber : 212-279-8