Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bromomaleic anhydride, 97%, Acros Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
5926-51-2
C4HBrO3
176.95
MFCD00005519
YPRMWCKXOZFJGF-UHFFFAOYSA-N
bromomaleic anhydride,2,5-furandione, 3-bromo,3-bromo-2,5-furandione,bromomaleicanhydride,monobromomaleic anhydride,3-bromo-2,5-furanedione,2,5-furandione,3-bromo,2-bromomaleic acid anhydride,3-bromo-2,5-furandione #
80027
3-bromofuran-2,5-dione
C1=C(C(=O)OC1=O)Br
5926-51-2
C4HBrO3
176.95
MFCD00005519
YPRMWCKXOZFJGF-UHFFFAOYSA-N
bromomaleic anhydride,2,5-furandione, 3-bromo,3-bromo-2,5-furandione,bromomaleicanhydride,monobromomaleic anhydride,3-bromo-2,5-furanedione,2,5-furandione,3-bromo,2-bromomaleic acid anhydride,3-bromo-2,5-furandione #
80027
3-bromofuran-2,5-dione
C1=C(C(=O)OC1=O)Br
Quantity 50g
Packaging Glass bottle
Specific Gravity 1.905
Flash Point 113°C
Refractive Index 1.54
Boiling Point 215.0°C
Density 1.9050g/mL
Assay Percent Range 97%
Beilstein 17, 435
Formula Weight 176.95
Percent Purity 97%
Chemical Name or Material Bromomaleic anhydride
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger