Alfa Aesar™ Bromomalonaldehyde, 97%

Manufacturer: Alfa Aesar™ H3103003

Quantity 1g

 View more versions of this product

Catalog No. AAH3103003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2065-75-0
    Molecular Formula C3H3BrO2
    Linear Formula BrCH(CHO)2
    Formula Weight 150.96
    MDL Number MFCD00459999
    Chemical Name or Material Bromomalonaldehyde

    Specifications

    Formula Weight 150.96
    Melting Point ≈145°C
    Quantity 1g
    Molecular Formula C3H3BrO2
    Linear Formula BrCH(CHO)2
    MDL Number MFCD00459999
    CAS 2065-75-0
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280i-P305+P351+P338
    Wear eye/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Chemical Name or Material Bromomalonaldehyde
    Assay 97%

    GHS H Statement
    H318-H302
    Causes serious eye damage.
    Harmful if swallowed.

    GHS P Statement
    P280i-P305+P351+P338
    Wear eye/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit